Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696393.3 | XP_005696450.2 | 8 - 9 | 1761 | 79 | |
| XM_005696394.3 | XP_005696451.2 | 8 - 9 | 1756 | 79 | ||
| XM_005696395.3 | XP_005696452.2 | 8 - 9 | 1802 | 79 | ||
| XM_013973872.2 | XP_013829326.2 | 8 - 9 | 1893 | 79 | ||
| GenBank mRNA | JO637931.2 | - | 5 - 6 | 843 | 79 | |
More Information
Set Membership: |
|

MMUT
Genomic Map