Assay Details
| Gene Symbol: LOC102188106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698181.2 | XP_005698238.2 | 7 - 8 | 868 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102188106| Gene Symbol: LOC102188106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698181.2 | XP_005698238.2 | 7 - 8 | 868 | 77 | |
Set Membership: |
|