Assay Details
Gene Symbol: LOC102186671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045147.1 | XP_017900636.1 | 1846 | 103 | ||
GenBank mRNA | JO587095.2 | - | 1548 | 103 |

More Information
Set Membership: |
![]() ![]() |
