Assay Details
| Gene Symbol: LOC102186671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045147.1 | XP_017900636.1 | 2655 | 99 | ||
| GenBank mRNA | JO587095.2 | - | 2357 | 99 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102186671| Gene Symbol: LOC102186671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045147.1 | XP_017900636.1 | 2655 | 99 | ||
| GenBank mRNA | JO587095.2 | - | 2357 | 99 | ||