Assay Details
Gene Symbol: LEPROT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045213.1 | XP_017900702.1 | 2 - 3 | 189 | 67 | |
XM_018045217.1 | XP_017900706.1 | 2 - 3 | 189 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
