Assay Details
| Gene Symbol: PLA2G12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964470.2 | XP_013819924.1 | 1 - 2 | 380 | 66 | |
| XM_018049225.1 | XP_017904714.1 | 1 - 2 | 391 | 66 | ||
More Information
Set Membership: |
|

PLA2G12A
Genomic Map