Assay Details
Gene Symbol: C7H1orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051585.1 | XP_017907074.1 | 6 - 7 | 866 | 95 | |
XM_018051586.1 | XP_017907075.1 | 5 - 6 | 664 | 95 | ||
GenBank mRNA | KA308604.1 | - | 5 - 6 | 770 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
