Assay Details
Gene Symbol: C7H1orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051585.1 | XP_017907074.1 | 8 - 9 | 1080 | 83 | |
XM_018051586.1 | XP_017907075.1 | 7 - 8 | 878 | 83 | ||
GenBank mRNA | KA308604.1 | - | 7 - 8 | 984 | 83 |
Gene Symbol: CCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO429302.2 | - | 2024 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
