Assay Details
Gene Symbol: SP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064790.1 | XP_017920279.1 | 5 - 6 | 2371 | 86 | |
XM_018064800.1 | XP_017920289.1 | 5 - 6 | 1796 | 86 | ||
XM_018064805.1 | XP_017920294.1 | 4 - 5 | 1784 | 86 | ||
GenBank mRNA | JO339810.2 | - | 2 - 3 | 1124 | 86 | |
JO428667.2 | - | 2 - 3 | 708 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
