Assay Details
| Gene Symbol: SLC9A3R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972188.2 | XP_013827642.2 | 4 - 5 | 1064 | 62 | |
| GenBank mRNA | JO631760.2 | - | 4 - 5 | 1094 | 62 | |
More Information
Set Membership: |
|

SLC9A3R1
Genomic Map