Assay Details
Gene Symbol: APOPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066189.1 | XP_017921678.1 | 3 - 4 | 641 | 56 | |
GenBank mRNA | KA338927.1 | - | 3 - 4 | 459 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
