Assay Details
| Gene Symbol: IGSF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675436.3 | XP_005675493.2 | 5 - 6 | 5529 | 74 | |
| XM_018048958.1 | XP_017904447.1 | 6 - 7 | 5727 | 74 | ||
| XM_018048968.1 | XP_017904457.1 | 7 - 8 | 5604 | 74 | ||
| XM_018048977.1 | XP_017904466.1 | 6 - 7 | 5607 | 74 | ||
| XM_018048993.1 | XP_017904482.1 | 6 - 7 | 5604 | 74 | ||
| XM_018049001.1 | XP_017904490.1 | 5 - 6 | 5285 | 74 | ||
More Information
Set Membership: |
|

IGSF10
Genomic Map