Assay Details
Gene Symbol: ACPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675609.3 | XP_005675666.1 | 4 - 5 | 553 | 69 | |
XM_005675610.2 | XP_005675667.1 | 4 - 5 | 553 | 69 | ||
XM_018051351.1 | XP_017906840.1 | 4 - 5 | 553 | 69 | ||
GenBank mRNA | KA349165.1 | - | 7 - 8 | 1326 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
