Assay Details
Gene Symbol: OLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680848.3 | XP_005680905.1 | 4 - 5 | 624 | 76 | |
XM_005680849.3 | XP_005680906.1 | 4 - 5 | 615 | 76 | ||
GenBank mRNA | JO565841.2 | - | 2 - 3 | 186 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
