Assay Details
| Gene Symbol: N4BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050581.1 | XP_017906070.1 | 4 - 5 | 1411 | 58 | |
| XM_018050582.1 | XP_017906071.1 | 4 - 5 | 1354 | 58 | ||
| XM_018050583.1 | XP_017906072.1 | 4 - 5 | 1409 | 58 | ||
| GenBank mRNA | KA351871.1 | - | 4 - 5 | 1378 | 58 | |
More Information
Set Membership: |
|

N4BP3
Genomic Map