Assay Details
| Gene Symbol: CABP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061004.1 | XP_017916493.1 | 2 - 3 | 269 | 77 | |
| XM_018061005.1 | XP_017916494.1 | 2 - 3 | 269 | 77 | ||
More Information
Set Membership: |
|

CABP7
Genomic Map