Assay Details
Gene Symbol: TC2N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973205.2 | XP_013828659.1 | 6 - 7 | 676 | 67 | |
XM_018066464.1 | XP_017921953.1 | 7 - 8 | 883 | 67 | ||
XM_018066465.1 | XP_017921954.1 | 7 - 8 | 921 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
