Assay Details
| Gene Symbol: PRSS45P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066737.1 | XP_017922226.1 | 5 - 6 | 771 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS45P| Gene Symbol: PRSS45P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066737.1 | XP_017922226.1 | 5 - 6 | 771 | 90 | |
Set Membership: |
|