Assay Details
| Gene Symbol: C24H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697174.3 | XP_005697231.1 | 2 - 3 | 894 | 78 | |
| XM_018039570.1 | XP_017895059.1 | 2 - 3 | 1080 | 78 | ||
| XM_018039571.1 | XP_017895060.1 | 2 - 3 | 1080 | 78 | ||
More Information
Set Membership: |
|

C24H18orf25
Genomic Map