Assay Details
| Gene Symbol: C24H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697174.3 | XP_005697231.1 | 3 - 4 | 1080 | 64 | |
| XM_018039570.1 | XP_017895059.1 | 3 - 4 | 1266 | 64 | ||
| XM_018039571.1 | XP_017895060.1 | 3 - 4 | 1266 | 64 | ||
More Information
Set Membership: |
|

C24H18orf25
Genomic Map