Assay Details
| Gene Symbol: C24H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697174.3 | XP_005697231.1 | 4 - 5 | 1203 | 133 | |
| XM_018039570.1 | XP_017895059.1 | 4 - 5 | 1389 | 133 | ||
| XM_018039572.1 | XP_017895061.1 | 3 - 4 | 1226 | 133 | ||
| GenBank mRNA | JO433991.2 | - | 3 - 4 | 1206 | 133 | |
More Information
Set Membership: |
|

C24H18orf25
Genomic Map