Assay Details
| Gene Symbol: CHST12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041007.1 | XP_017896496.1 | 2 | 890 | 62 | |
| XM_018041008.1 | XP_017896497.1 | 2 | 885 | 62 | ||
| GenBank mRNA | JO619448.2 | - | 1 | 487 | 62 | |
More Information
Set Membership: |
|

CHST12
Genomic Map