Assay Details
Gene Symbol: HTATIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043525.1 | XP_017899014.1 | 2 - 3 | 382 | 71 | |
GenBank mRNA | JO568773.2 | - | 2 - 3 | 363 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
