Assay Details
| Gene Symbol: RALY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057731.1 | XP_017913220.1 | 3 - 4 | 810 | 75 | |
| XM_018057732.1 | XP_017913221.1 | 2 - 3 | 727 | 75 | ||
| XM_018057733.1 | XP_017913222.1 | 3 - 4 | 810 | 75 | ||
More Information
Set Membership: |
|

RALY
Genomic Map