Assay Details
| Gene Symbol: RMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039478.1 | XP_017894967.1 | 17 - 18 | 1743 | 74 | |
| GenBank mRNA | JO638954.2 | - | 3 - 4 | 412 | 74 | |
| JT685598.1 | - | 8 - 9 | 1723 | 74 | ||
More Information
Set Membership: |
|

RMC1
Genomic Map