Assay Details
Gene Symbol: STN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041596.1 | XP_017897085.1 | 5 - 6 | 725 | 64 | |
XM_018041597.1 | XP_017897086.1 | 5 - 6 | 724 | 64 | ||
XM_018041598.1 | XP_017897087.1 | 5 - 6 | 675 | 64 | ||
XM_018041599.1 | XP_017897088.1 | 5 - 6 | 541 | 64 | ||
XM_018041600.1 | XP_017897089.1 | 5 - 6 | 675 | 64 | ||
XM_018041601.1 | XP_017897090.1 | 5 - 6 | 726 | 64 | ||
GenBank mRNA | JO353129.2 | - | 2 - 3 | 247 | 64 | |
JO578636.1 | - | 7 - 8 | 1014 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
