Assay Details
| Gene Symbol: STOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041957.1 | XP_017897446.1 | 3 - 4 | 706 | 84 | |
| XM_018041958.1 | XP_017897447.1 | 3 - 4 | 706 | 84 | ||
| XM_018041959.1 | XP_017897448.1 | 3 - 4 | 706 | 84 | ||
| XM_018041961.1 | XP_017897450.1 | 2 - 3 | 2110 | 84 | ||
| XM_018041962.1 | XP_017897451.1 | 2 - 3 | 703 | 84 | ||
| GenBank mRNA | KA329989.1 | - | 1 - 2 | 1135 | 84 | |
More Information
Set Membership: |
|

STOX2
Genomic Map