Assay Details
| Gene Symbol: NIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677175.2 | XP_005677232.1 | 3 - 4 | 450 | 67 | |
| XM_005677176.3 | XP_005677233.1 | 2 - 3 | 570 | 67 | ||
| GenBank mRNA | JO426840.1 | - | 2509 | 67 | ||
More Information
Set Membership: |
|

NIT1
Genomic Map