Assay Details
| Gene Symbol: LOC102180455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047122.1 | XP_017902611.1 | 17 - 18 | 2051 | 86 | |
| XM_018047124.1 | XP_017902613.1 | 16 - 17 | 2193 | 86 | ||
| GenBank mRNA | JO618351.2 | - | 2 - 3 | 225 | 86 | |
More Information
Set Membership: |
|

LOC102180455
Genomic Map