Assay Details
| Gene Symbol: LOC102180455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963332.2 | XP_013818786.2 | 4 - 5 | 727 | 79 | |
| XM_018047122.1 | XP_017902611.1 | 5 - 6 | 585 | 79 | ||
| XM_018047123.1 | XP_017902612.1 | 4 - 5 | 727 | 79 | ||
| XM_018047124.1 | XP_017902613.1 | 4 - 5 | 727 | 79 | ||
More Information
Set Membership: |
|

LOC102180455
Genomic Map