Assay Details
Gene Symbol: ATF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679967.3 | XP_005680024.1 | 6 - 7 | 836 | 65 | |
XM_005679969.3 | XP_005680026.1 | 5 - 6 | 714 | 65 | ||
XM_013963808.2 | XP_013819262.1 | 6 - 7 | 870 | 65 | ||
XM_018047784.1 | XP_017903273.1 | 6 - 7 | 806 | 65 | ||
XM_018047785.1 | XP_017903274.1 | 6 - 7 | 808 | 65 | ||
XM_018047787.1 | XP_017903276.1 | 6 - 7 | 913 | 65 | ||
XM_018047788.1 | XP_017903277.1 | 6 - 7 | 1057 | 65 | ||
XM_018047789.1 | XP_017903278.1 | 4 - 5 | 553 | 65 | ||
XM_018047790.1 | XP_017903279.1 | 5 - 6 | 801 | 65 | ||
XM_018047791.1 | XP_017903280.1 | 5 - 6 | 932 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
