Assay Details
Gene Symbol: MED10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065763.1 | XP_017921252.1 | 2 - 3 | 282 | 86 | |
GenBank mRNA | JO641623.2 | - | 2 - 3 | 535 | 86 | |
JT695507.1 | - | 3 - 4 | 594 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
