Assay Details
| Gene Symbol: EBI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682564.3 | XP_005682621.1 | 1 - 2 | 1027 | 58 | |
| GenBank mRNA | JO598143.2 | - | 1 - 2 | 198 | 58 | |
More Information
Set Membership: |
|

EBI3
Genomic Map