Assay Details
| Gene Symbol: GLRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061476.1 | XP_017916965.1 | 8 - 9 | 1153 | 57 | |
| GenBank mRNA | JO643860.2 | - | 2 - 3 | 1048 | 57 | |
More Information
Set Membership: |
|

GLRB
Genomic Map