Assay Details
| Gene Symbol: SHOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698466.3 | XP_005698523.1 | 5 - 6 | 1478 | 86 | |
| XM_005698467.3 | XP_005698524.1 | 5 - 6 | 1469 | 86 | ||
| XM_018041275.1 | XP_017896764.1 | 5 - 6 | 1464 | 86 | ||
| XM_018041276.1 | XP_017896765.1 | 5 - 6 | 1567 | 86 | ||
More Information
Set Membership: |
|

SHOC2
Genomic Map