Assay Details
| Gene Symbol: CHCHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699218.3 | XP_005699275.1 | 1 - 2 | 147 | 89 | |
| GenBank mRNA | JI861776.1 | - | 1 - 2 | 121 | 89 | |
| JO573406.2 | - | 2 - 3 | 427 | 89 | ||
More Information
Set Membership: |
|

CHCHD1
Genomic Map