Assay Details
Gene Symbol: TMEM35B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041256.1 | XP_017896745.1 | 2 - 3 | 1036 | 95 | |
GenBank mRNA | JO602556.2 | - | 2 - 3 | 658 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
