Assay Details
Gene Symbol: EVA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047722.1 | XP_017903211.1 | 7 - 8 | 1353 | 52 | |
XM_018047783.1 | XP_017903272.1 | 5 - 6 | 1022 | 52 | ||
GenBank mRNA | JO366538.2 | - | 4 - 5 | 220 | 52 | |
JO451785.2 | - | 1 - 2 | 509 | 52 | ||
JO518325.2 | - | 1 - 2 | 405 | 52 | ||
JO615093.2 | - | 1 - 2 | 535 | 52 | ||
JO618865.2 | - | 1 - 2 | 539 | 52 | ||
JO618866.2 | - | 1 - 2 | 539 | 52 | ||
JT705608.1 | - | 2 - 3 | 434 | 52 | ||
JT717665.1 | - | 1 - 2 | 216 | 52 | ||
KA340698.1 | - | 2 - 3 | 245 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
