Assay Details
| Gene Symbol: TINAGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057412.1 | XP_017912901.1 | 7 - 8 | 967 | 55 | |
| XM_018057419.1 | XP_017912908.1 | 7 - 8 | 1059 | 55 | ||
| XM_018057425.1 | XP_017912914.1 | 7 - 8 | 1088 | 55 | ||
| GenBank mRNA | JO375541.2 | - | 5 - 6 | 773 | 55 | |
More Information
Set Membership: |
|

TINAGL1
Genomic Map