Assay Details
| Gene Symbol: AK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045182.1 | XP_017900671.1 | 5 - 6 | 779 | 69 | |
| XM_018045191.1 | XP_017900680.1 | 5 - 6 | 790 | 69 | ||
| GenBank mRNA | JT664494.1 | - | 6 - 7 | 3026 | 69 | |
More Information
Set Membership: |
|

AK4
Genomic Map