Assay Details
Gene Symbol: ADCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679526.3 | XP_005679583.2 | 6 - 7 | 1683 | 63 | |
GenBank mRNA | JO592296.2 | - | 2 - 3 | 468 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
