Assay Details
| Gene Symbol: ATP6V1G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684344.3 | XP_005684401.1 | 2 - 3 | 319 | 89 | |
| GenBank mRNA | JO413238.2 | - | 1 - 2 | 502 | 89 | |
| JO446638.2 | - | 1 - 2 | 490 | 89 | ||
More Information
Set Membership: |
|

ATP6V1G1
Genomic Map