Assay Details
| Gene Symbol: C15H11orf42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689772.3 | XP_005689829.2 | 1 - 2 | 3384 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C15H11orf42| Gene Symbol: C15H11orf42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689772.3 | XP_005689829.2 | 1 - 2 | 3384 | 81 | |
Set Membership: |
|