Assay Details
Gene Symbol: NAT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972189.2 | XP_013827643.1 | 3 - 4 | 294 | 84 | |
XM_018063861.1 | XP_017919350.1 | 4 - 5 | 431 | 84 | ||
XM_018063862.1 | XP_017919351.1 | 4 - 5 | 413 | 84 | ||
XM_018063863.1 | XP_017919352.1 | 3 - 4 | 335 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
