Assay Details
| Gene Symbol: NQO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039122.1 | XP_017894611.1 | 3 - 4 | 476 | 65 | |
| XM_018039123.1 | XP_017894612.1 | 4 - 5 | 526 | 65 | ||
| XM_018039124.1 | XP_017894613.1 | 4 - 5 | 523 | 65 | ||
| XM_018039125.1 | XP_017894614.1 | 3 - 4 | 486 | 65 | ||
| XM_018039126.1 | XP_017894615.1 | 3 - 4 | 480 | 65 | ||
| GenBank mRNA | JO412229.2 | - | 4 - 5 | 720 | 65 | |
| JO595790.2 | - | 3 - 4 | 512 | 65 | ||
More Information
Set Membership: |
|

NQO2
Genomic Map