Assay Details
| Gene Symbol: LOC102169925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039758.1 | XP_017895247.1 | 5 - 6 | 941 | 100 | |
| XM_018039759.1 | XP_017895248.1 | 6 - 7 | 823 | 100 | ||
| GenBank mRNA | JO584338.1 | - | 5 - 6 | 687 | 100 | |
More Information
Set Membership: |
|

LOC102169925
Genomic Map