Assay Details
Gene Symbol: DCTPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041031.1 | XP_017896520.1 | 2 - 3 | 316 | 96 | |
GenBank mRNA | JO587331.2 | - | 2 - 3 | 266 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
