Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042570.1 | XP_017898059.1 | 4 - 5 | 1716 | 85 | |
XM_018042572.1 | XP_017898061.1 | 3 - 4 | 1692 | 85 | ||
XM_018042573.1 | XP_017898062.1 | 4 - 5 | 1716 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
