Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042570.1 | XP_017898059.1 | 9 - 10 | 2588 | 90 | |
XM_018042572.1 | XP_017898061.1 | 8 - 9 | 2564 | 90 | ||
XM_018042573.1 | XP_017898062.1 | 9 - 10 | 2438 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
