Assay Details
Gene Symbol: SLC15A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043203.1 | XP_017898692.1 | 3 - 4 | 1036 | 65 | |
GenBank mRNA | JO623153.2 | - | 3 - 4 | 1049 | 65 | |
JT668566.1 | - | 7 - 8 | 2198 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
